Learn More
ACROBiosystems Recombinant Protein;FITC-Labeled Human BCMA / TNFRSF17 Protein, His Tag;HEK293;25UG;BCA,TNFRSF17,CD269,BCM,BCMA,TNFRSF-17
Catalog No. 501657454
Encompass_Preferred
Recombinant Protein;FITC-Labeled Human BCMA / TNFRSF17 Protein, His Tag;25UG;FITC-Labeled Human BCMA, His Tag (Cat. No. BCA-HF2H1) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession # Q02223-1). It is the FITC labeled form of Human BCMA, His Tag (Cat. No. BCA-H522y).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.8 kDa. The protein migrates as 10 kDa and 14-16 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;293 cells were transfected with anti-BCMA-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-Labeled Human BCMA, His Tag (Cat. No. BCA-HF2H1, 1 µg/ml) and C. FITC-labeled protein control. A. Non-transfected 293 cells and C. FITC-labeled protein control were used as negative control. RFP was used to evaluate CAR (anti-BCMA-scFv) expression and FITC was used to evaluate the binding activity of FITC-Labeled Human BCMA, His Tag (Cat. No. BCA-HF2H1) (QC tested).
Catalog No. 50-165-7454 Supplier ACROBiosystems Supplier No. BCAHF2H125UG
Recombinant Protein;FITC-Labeled Human BCMA / TNFRSF17 Protein, His Tag;25UG;FITC-Labeled Human BCMA, His Tag (Cat. No. BCA-HF2H1) is expressed from human HEK293 cells. It contains AA Met 1 - Ala 54 (Accession # Q02223-1). It is the FITC labeled form of Human BCMA, His Tag (Cat. No. BCA-H522y).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.8 kDa. The protein migrates as 10 kDa and 14-16 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;293 cells were transfected with anti-BCMA-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-Labeled Human BCMA, His Tag (Cat. No. BCA-HF2H1, 1 µg/ml) and C. FITC-labeled protein control. A. Non-transfected 293 cells and C. FITC-labeled protein control were used as negative control. RFP was used to evaluate CAR (anti-BCMA-scFv) expression and FITC was used to evaluate the binding activity of FITC-Labeled Human BCMA, His Tag (Cat. No. BCA-HF2H1) (QC tested).