Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human BCMA / TNFRSF17 Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;BCA,TNFRSF17,CD269,BCM,BCMA,TNFRSF-17
Catalog No. 501658018
Encompass_Preferred
Recombinant Protein;Biotinylated Human BCMA / TNFRSF17 Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human BCMA, His Tag, Avi Tag (BCA-H82E4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 9.6 kDa. The protein migrates as 12 kDa and 15-17 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BCMA, His Tag, Avi Tag (Cat. No. BCA-H82E4) with a linear range of 2-31 ng/mL (QC tested).;Immobilized Anti-Human BCMA MAb at 2 μg/mL (100 μL/well) can bind Biotinylated Human BCMA, His Tag, Avi Tag (Cat. No. BCA-H82E4) with a linear range of 0.5-8 ng/mL (Routinely tested).;293 cells were transfected with anti-BCMA-scFv and RFP tag. 2e5 of the cells were first stained with B. Biotinylated Huma
Catalog No. 50-165-8018 Supplier ACROBiosystems Supplier No. BCAH82E4200UG
Recombinant Protein;Biotinylated Human BCMA / TNFRSF17 Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human BCMA, His Tag, Avi Tag (BCA-H82E4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 9.6 kDa. The protein migrates as 12 kDa and 15-17 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BCMA, His Tag, Avi Tag (Cat. No. BCA-H82E4) with a linear range of 2-31 ng/mL (QC tested).;Immobilized Anti-Human BCMA MAb at 2 μg/mL (100 μL/well) can bind Biotinylated Human BCMA, His Tag, Avi Tag (Cat. No. BCA-H82E4) with a linear range of 0.5-8 ng/mL (Routinely tested).;293 cells were transfected with anti-BCMA-scFv and RFP tag. 2e5 of the cells were first stained with B. Biotinylated Huma