Learn More
ACROBiosystems Recombinant Protein;Human BCMA / TNFRSF17 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;BCA,TNFRSF17,CD269,BCM,BCMA,TNFRSF-17
Catalog No. 501657575
Encompass_Preferred
Recombinant Protein;Human BCMA / TNFRSF17 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human BCMA, Mouse IgG2a Fc Tag, low endotoxin (BCA-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 32.8 kDa. The protein migrates as 35-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 2 μg/mL (100 μL/well) can bind Human BCMA, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. BCA-H5253) with a linear range of 0.2-0.8 ng/mL (QC tested).;Immobilized Human BCMA, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. BCA-H5253) at 2 μg/mL (100 μL/well) can bind Anti-Human BCMA Mab with a linear range of 0.2-3 ng/mL (Routinely tested).
Catalog No. 50-165-7575 Supplier ACROBiosystems Supplier No. BCAH5253100UG
Recombinant Protein;Human BCMA / TNFRSF17 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human BCMA, Mouse IgG2a Fc Tag, low endotoxin (BCA-H5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 32.8 kDa. The protein migrates as 35-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag (Cat. No. BAF-H4268) at 2 μg/mL (100 μL/well) can bind Human BCMA, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. BCA-H5253) with a linear range of 0.2-0.8 ng/mL (QC tested).;Immobilized Human BCMA, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. BCA-H5253) at 2 μg/mL (100 μL/well) can bind Anti-Human BCMA Mab with a linear range of 0.2-3 ng/mL (Routinely tested).