Learn More
Aviva Systems Biology Corp BCO1 Antibody - C-terminal region (ARP64136_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64136_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCMO1. It was validated on Western Blot. . Alternate names: BCDO, BCDO1, BCMO, BCO, BCO1, FLJ10730ApplicationWB. Species reactivity: Human
Catalog No. 50-288-182