Learn More
ABclonal Technology BCMO1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A158481000UL
Encompass_Preferred
Vitamin A metabolism is important for vital processes such as vision, embryonic development, cell differentiation, and membrane and skin protection. The protein encoded by this gene is a key enzyme in beta-carotene metabolism to vitamin A. It catalyzes the oxidative cleavage of beta,beta-carotene into two retinal molecules.
Catalog No. 50-001-08741