Learn More
Medchemexpress LLC BCN-endo-PEG4-NHS 50mg
SDP
Catalog No. 5000205963
Encompass_Preferred
BCN-endo-PEG4-NHS is an ADC Linker containing 4 PEG units BCN-endo-PEG4-NHS contains the lyophilic bidentate macrocyclic ligand endo-BCN which allows for further synthesis of macrocyclic complexes In click chemistry endo-BCN can react with molecules containing azide groups to form stable triazoles in the absence of catalysts
Catalog No. 50-002-05963 Supplier Medchemexpress LLC Supplier No. HYW09607950MG
BCN-endo-PEG4-NHS is an ADC Linker containing 4 PEG units BCN-endo-PEG4-NHS contains the lyophilic bidentate macrocyclic ligand endo-BCN which allows for further synthesis of macrocyclic complexes In click chemistry endo-BCN can react with molecules containing azide groups to form stable triazoles in the absence of catalysts