Learn More
Medchemexpress LLC Bcn-endo-PEG7-maleimide | 95.0% | 695.80 g/mol | C34H53N3O12 | 10 MG
SDP
Catalog No. 5000204363
Encompass_Preferred
BCN-endo-PEG7-maleimide is a BCN-based click chemistry linker containing a seven-unit polyethylene glycol (PEG7) spacer and a terminal maleimide for selective thiol conjugation. It is designed for bioconjugation, antibody-drug conjugate linker assembly, and other strain-promoted click chemistry applications.
  • Contains an endo-BCN moiety for strain-promoted click reactions.
  • Includes a PEG7 spacer to improve solubility and flexibility.
  • Has a terminal maleimide for selective conjugation to thiol groups.
  • Suitable for bioconjugation and antibody-drug conjugate linker synthesis.
  • Offered in small-scale research pack sizes for discovery workflows.
Catalog No. 50-002-04363 Supplier Medchemexpress LLC Supplier No. HY15631710MG
BCN-endo-PEG7-maleimide is a BCN-based click chemistry linker containing a seven-unit polyethylene glycol (PEG7) spacer and a terminal maleimide for selective thiol conjugation. It is designed for bioconjugation, antibody-drug conjugate linker assembly, and other strain-promoted click chemistry applications.
  • Contains an endo-BCN moiety for strain-promoted click reactions.
  • Includes a PEG7 spacer to improve solubility and flexibility.
  • Has a terminal maleimide for selective conjugation to thiol groups.
  • Suitable for bioconjugation and antibody-drug conjugate linker synthesis.
  • Offered in small-scale research pack sizes for discovery workflows.