Learn More
Medchemexpress LLC BCN-exo-PEG3-NH2 | 1841134-72-2 | >95.0% | C19H32N2O5 | 10MG
SDP
Catalog No. 5000260525
Encompass_Preferred
BCN-exo-PEG3-NH2 is a BCN-functionalized PEG3 amine linker used in bioorthogonal conjugation and PROTAC linker synthesis. It provides a strained alkyne handle and a terminal primary amine to enable modular coupling strategies, and is commonly supplied in research-scale quantities at high reported purity.
  • Provides a strained alkyne (BCN) for rapid bioorthogonal click reactions.
  • Contains a PEG3 spacer for flexibility and improved solubility.
  • Features a terminal primary amine for amide or reductive amination conjugation.
  • Suitable for linker construction in targeted degradation and bioconjugation workflows.
  • Supplied in research-scale quantities with supplier-reported high purity.
Catalog No. 50-002-60525 Supplier Medchemexpress LLC Supplier No. HY13340210MG
BCN-exo-PEG3-NH2 is a BCN-functionalized PEG3 amine linker used in bioorthogonal conjugation and PROTAC linker synthesis. It provides a strained alkyne handle and a terminal primary amine to enable modular coupling strategies, and is commonly supplied in research-scale quantities at high reported purity.
  • Provides a strained alkyne (BCN) for rapid bioorthogonal click reactions.
  • Contains a PEG3 spacer for flexibility and improved solubility.
  • Features a terminal primary amine for amide or reductive amination conjugation.
  • Suitable for linker construction in targeted degradation and bioconjugation workflows.
  • Supplied in research-scale quantities with supplier-reported high purity.