Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BCN-exo-PEG3-NH2 | 1841134-72-2 | 99.9% | 368.47 | 250 MG
SDP

Catalog No. 5000366101
Encompass_Preferred

BCN-exo-PEG3-NH2 is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring a BCN group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Purity: 99.94%
  • Molecular weight: 368.47
  • Chemical formula: C19H32N2O5
  • CAS number: 1841134-72-2
  • Appearance: Colorless to light yellow liquid
  • Density: 1.14±0.1 g/cm3
  • Used in PROTAC synthesis
  • Contains a BCN group for SPAAC reactions

Catalog No. 50-003-66101 Supplier Medchemexpress LLC Supplier No. 250MGHY133402
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BCN-exo-PEG3-NH2 is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring a BCN group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Purity: 99.94%
  • Molecular weight: 368.47
  • Chemical formula: C19H32N2O5
  • CAS number: 1841134-72-2
  • Appearance: Colorless to light yellow liquid
  • Density: 1.14±0.1 g/cm3
  • Used in PROTAC synthesis
  • Contains a BCN group for SPAAC reactions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.