Learn More
Medchemexpress LLC BCN-exo-PEG7-maleimide | 2143965-47-1 | 95.1% | 695.80 | C34H53N3O12 | 5 MG
SDP
Catalog No. 5000221872
Encompass_Preferred
BCN-exo-PEG7-maleimide is an ADC linker that combines a bicyclononyne (BCN) alkyne for strain-promoted azide-alkyne cycloaddition with a thiol-reactive maleimide, joined by a seven-unit polyethylene glycol spacer. It is designed for catalyst-free click conjugation and thiol-specific bioconjugation in ADC construction and biomolecule labeling applications.
  • Enables catalyst-free azide-alkyne cycloaddition (SPAAC) via BCN.
  • Provides thiol-selective conjugation through maleimide chemistry.
  • Includes a PEG7 spacer to improve solubility and reduce steric hindrance.
  • Suitable for antibody-drug conjugate assembly and general bioconjugation workflows.
  • Supplied in research-scale quantities with batch-specific certificate of analysis.
Catalog No. 50-002-21872 Supplier Medchemexpress LLC Supplier No. HY1347345MG
BCN-exo-PEG7-maleimide is an ADC linker that combines a bicyclononyne (BCN) alkyne for strain-promoted azide-alkyne cycloaddition with a thiol-reactive maleimide, joined by a seven-unit polyethylene glycol spacer. It is designed for catalyst-free click conjugation and thiol-specific bioconjugation in ADC construction and biomolecule labeling applications.
  • Enables catalyst-free azide-alkyne cycloaddition (SPAAC) via BCN.
  • Provides thiol-selective conjugation through maleimide chemistry.
  • Includes a PEG7 spacer to improve solubility and reduce steric hindrance.
  • Suitable for antibody-drug conjugate assembly and general bioconjugation workflows.
  • Supplied in research-scale quantities with batch-specific certificate of analysis.