Learn More
Medchemexpress LLC Bcn-PEG1-Val-Cit-PABC-OH | 95.0% | 670.80 g/mol | C34H50N6O8 | 5 MG
SDP
Catalog No. 5000203390
Encompass_Preferred
BCN-PEG1-Val-Cit-PABC-OH is a BCN-functionalized, cleavable PEG-based antibody-drug conjugate (ADC) linker containing a Val-Cit dipeptide and a para-aminobenzyl carbamate self-immolative spacer. It is supplied as a powder for conjugation and click chemistry with azide-functionalized payloads.
  • Cleavable Val-Cit dipeptide with PABC self-immolative spacer.
  • Single-unit PEG (PEG1) provides solubility and spacing.
  • BCN group enables strain-promoted azide-alkyne cycloaddition (SPAAC) conjugation.
  • Supplied as powder, soluble in DMSO, store at -20°C.
  • Purity typically 95.0% and molecular weight ~670.80 g/mol.
Catalog No. 50-002-03390 Supplier Medchemexpress LLC Supplier No. HY1309235MG
BCN-PEG1-Val-Cit-PABC-OH is a BCN-functionalized, cleavable PEG-based antibody-drug conjugate (ADC) linker containing a Val-Cit dipeptide and a para-aminobenzyl carbamate self-immolative spacer. It is supplied as a powder for conjugation and click chemistry with azide-functionalized payloads.
  • Cleavable Val-Cit dipeptide with PABC self-immolative spacer.
  • Single-unit PEG (PEG1) provides solubility and spacing.
  • BCN group enables strain-promoted azide-alkyne cycloaddition (SPAAC) conjugation.
  • Supplied as powder, soluble in DMSO, store at -20°C.
  • Purity typically 95.0% and molecular weight ~670.80 g/mol.