Learn More
Medchemexpress LLC BCN-PEG3-biotin | 1263166-92-2 | 95.0% | 594.76 | C29H46N4O7S | 100 MG
SDP
Catalog No. 5000201884
Encompass_Preferred
BCN-PEG3-biotin is a click-chemistry linker containing a bicyclononyne (BCN) group, a three-unit polyethylene glycol spacer, and a terminal biotin. It is used for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules and as a biotin tag for affinity capture or detection.
  • Facilitates copper-free SPAAC click reactions.
  • Three-unit PEG spacer improves solubility and reduces steric hindrance.
  • Terminal biotin enables streptavidin-based capture and detection.
  • Supplied as an off-white to light yellow solid with ≥95.0% purity.
  • Soluble in DMSO (100 mg/mL); powder storage recommended at -20°C.
Catalog No. 50-002-01884 Supplier Medchemexpress LLC Supplier No. HY130924100MG
BCN-PEG3-biotin is a click-chemistry linker containing a bicyclononyne (BCN) group, a three-unit polyethylene glycol spacer, and a terminal biotin. It is used for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules and as a biotin tag for affinity capture or detection.
  • Facilitates copper-free SPAAC click reactions.
  • Three-unit PEG spacer improves solubility and reduces steric hindrance.
  • Terminal biotin enables streptavidin-based capture and detection.
  • Supplied as an off-white to light yellow solid with ≥95.0% purity.
  • Soluble in DMSO (100 mg/mL); powder storage recommended at -20°C.