Learn More
Medchemexpress LLC Bcn-peg3-oh | 2126749-77-5 | 369.45 g/mol | C19H31NO6 | 100 MG
SDP
Catalog No. 5000204292
Encompass_Preferred
BCN-PEG3-OH is a BCN-functionalized PEG linker used for copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) and bioconjugation applications such as antibody-drug conjugate assembly. It provides a short PEG spacer with a reactive bicyclononyne moiety to enable site-selective conjugation without metal catalysts.
  • Bcn functional group enables copper-free SPAAC click chemistry.
  • Three-unit PEG spacer provides flexibility and solubility for conjugates.
  • Suitable for antibody-drug conjugate synthesis and site-specific bioconjugation.
  • Reactive alcohol terminus allows further functionalization or coupling reactions.
  • Compact molecular weight and defined formula support precise stoichiometry in conjugation workflows.
Catalog No. 50-002-04292 Supplier Medchemexpress LLC Supplier No. HY130925100MG
BCN-PEG3-OH is a BCN-functionalized PEG linker used for copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) and bioconjugation applications such as antibody-drug conjugate assembly. It provides a short PEG spacer with a reactive bicyclononyne moiety to enable site-selective conjugation without metal catalysts.
  • Bcn functional group enables copper-free SPAAC click chemistry.
  • Three-unit PEG spacer provides flexibility and solubility for conjugates.
  • Suitable for antibody-drug conjugate synthesis and site-specific bioconjugation.
  • Reactive alcohol terminus allows further functionalization or coupling reactions.
  • Compact molecular weight and defined formula support precise stoichiometry in conjugation workflows.