Learn More
Medchemexpress LLC Bcn-peg3-oh | 2126749-77-5 | 99.6% | 369.45 | C19H31NO6 | 10 MG
SDP
Catalog No. 5000202513
Encompass_Preferred
BCN-PEG3-OH is a BCN-functionalized PEG linker used as a non-cleavable linker and click-chemistry reagent for bioconjugation and antibody-drug conjugate synthesis. It features a BCN group for copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) and a three-unit PEG spacer terminating in a hydroxyl group.
  • Non-cleavable PEG linker for ADC synthesis
  • BCN group enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Three-unit PEG spacer with terminal hydroxyl
  • High purity suitable for research applications
  • Available in small pack sizes for laboratory use
Catalog No. 50-002-02513 Supplier Medchemexpress LLC Supplier No. HY13092510MG
BCN-PEG3-OH is a BCN-functionalized PEG linker used as a non-cleavable linker and click-chemistry reagent for bioconjugation and antibody-drug conjugate synthesis. It features a BCN group for copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) and a three-unit PEG spacer terminating in a hydroxyl group.
  • Non-cleavable PEG linker for ADC synthesis
  • BCN group enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Three-unit PEG spacer with terminal hydroxyl
  • High purity suitable for research applications
  • Available in small pack sizes for laboratory use