Learn More
Aviva Systems Biology Corp BCO2 Antibody - C-terminal region (ARP60586_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60586_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BCO2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human BCO2. Alternate names: B-DIOX-II, BCDO2, FLJ34464ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-287-093