Learn More
New England Biolabs, Inc. BcoDI – 1000 units
SDP
Catalog No. 504460
Encompass
A restriction endonuclease that recognizes the sequence GTCTCN^NNNN_.
Catalog No. 50-446-0 Supplier New England Biolabs, Inc. Supplier No. R0542S
A restriction endonuclease that recognizes the sequence GTCTCN^NNNN_.