Learn More
Molecular Research Center BCP
SDP
Catalog No. NC9551474
Encompass
BCP , 200 ml, a molecular biology grade 1-bromo-3-chloropropane.This new phase separation reagent substitutes for highly volatileand toxic chloroform in the TRI Reagent procedures
Catalog No. NC9551474 Supplier Molecular Research Center Supplier No. BP 151
Add to Cart
Add to Cart
BCP , 200 ml, a molecular biology grade 1-bromo-3-chloropropane.This new phase separation reagent substitutes for highly volatileand toxic chloroform in the TRI Reagent procedures