Learn More
Abnova™ bcr/abl DY Translocation FISH Probe
Catalog No. 89024807 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
Catalog No. Quantity
89-024-807 200 μL
1 options
Catalog No. 89-024-807 Supplier Abnova™ Supplier No. FT0002
Add to Cart
Add to Cart
Includes
DAPI Counterstain (1500ng/mL ) 250μL.
Labeled FISH probes for identification of gene translocation using Fluorescent In Situ Hybridization Technique.
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Content And Storage Store at 4°C in the dark.
For Use With (Application) Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
Includes DAPI Counterstain (1500 ng/mL ) 250 uL
Product Type bcr/abl DY Translocation FISH Probe
Quantity 200 μL