Learn More
GENEPATH DIAGNOSTICS INC BCR-ABL Major (p210) Quantification Control Material for qPCR with ABL Reference Gene--Std1-10^6 cps/µL, Std2-10^5 cps/µL, Std3-10^4 cps/µL, Std4-10^3 cps/µL, Std5-10^2 cps/µL, Std6-10^1 cps/µL (RUO)
Catalog No. NC3949369
Encompass_Preferred
The BCR-ABL Major (p210) Quantification Control Material is a validated reference standard designed for accurate quantitative measurement of BCR-ABL1 p210 fusion transcripts using real-time PCR (qPCR). The product includes calibrated BCR-ABL1 p210 along with ABL as a reference gene, enabling reliable normalization, calibration, and standard curve generation. This control material supports both absolute and relative quantification workflows and is suitable for use in assay development, validation, and routine quality control. It enables verification of assay performance parameters such as sensitivity, linearity, precision, and reproducibility, making it ideal for laboratories performing molecular monitoring of chronic myeloid leukemia (CML). Compatible with commonly used qPCR platforms and chemistries and integrates seamlessly into laboratory-developed test workflows. Designed for consistency and reproducibility ensuring reliable and comparable results across runs and instruments. (RUO)
Catalog No. NC3949369 Supplier GENEPATH DIAGNOSTICS INC Supplier No. 10830010050
The BCR-ABL Major (p210) Quantification Control Material is a validated reference standard designed for accurate quantitative measurement of BCR-ABL1 p210 fusion transcripts using real-time PCR (qPCR). The product includes calibrated BCR-ABL1 p210 along with ABL as a reference gene, enabling reliable normalization, calibration, and standard curve generation. This control material supports both absolute and relative quantification workflows and is suitable for use in assay development, validation, and routine quality control. It enables verification of assay performance parameters such as sensitivity, linearity, precision, and reproducibility, making it ideal for laboratories performing molecular monitoring of chronic myeloid leukemia (CML). Compatible with commonly used qPCR platforms and chemistries and integrates seamlessly into laboratory-developed test workflows. Designed for consistency and reproducibility ensuring reliable and comparable results across runs and instruments. (RUO)