Learn More
Medchemexpress LLC Bcr-abl1-in-1 | 1488090-21-6 | 99.8% | 359.30 | C18H12F3N3O2 | 5 MG
SDP
Catalog No. 5000221524
Encompass_Preferred
Bcr-abl1-in-1 is a small-molecule inhibitor targeting ABL1 (BCR-ABL1) for laboratory research. It shows potent activity with an IC50 of 8.7 nM against ABL1 (64-515), and is supplied as a high-purity solid suitable for in vitro biochemical and cell-based assays. This compound is intended for research use only and not for clinical or diagnostic applications.
  • Potent ABL1 inhibition with IC50 of 8.7 nM.
  • High reported purity (approximately 99.8%).
  • Molecular weight 359.30 and formula C18H12F3N3O2.
  • Supplied as a solid powder; soluble in DMSO for assays.
  • Suitable for in vitro biochemical and cell-based studies.
  • Intended for research use only, not for human use.
Catalog No. 50-002-21524 Supplier Medchemexpress LLC Supplier No. HY1458875MG
Bcr-abl1-in-1 is a small-molecule inhibitor targeting ABL1 (BCR-ABL1) for laboratory research. It shows potent activity with an IC50 of 8.7 nM against ABL1 (64-515), and is supplied as a high-purity solid suitable for in vitro biochemical and cell-based assays. This compound is intended for research use only and not for clinical or diagnostic applications.
  • Potent ABL1 inhibition with IC50 of 8.7 nM.
  • High reported purity (approximately 99.8%).
  • Molecular weight 359.30 and formula C18H12F3N3O2.
  • Supplied as a solid powder; soluble in DMSO for assays.
  • Suitable for in vitro biochemical and cell-based studies.
  • Intended for research use only, not for human use.