Learn More
Medchemexpress LLC Bcr-Abl1-In-1 | 1488090-21-6 | 99.82% | C18H12F3N3O2 | 100 MG
SDP
Catalog No. 5000354304
Encompass_Preferred
BCR-ABL1-IN-1 is an inhibitor for ABL1 (64-515) with an IC50 of 8.7 nM. It can be utilized in research concerning diseases related to the central nervous system (CNS).
  • Exhibits antiproliferative activity against mouse BaF3 cells expressing BCR-ABL with a GI50 of 69 nM and an IC50 of 75 nM.
  • Inhibits the proliferation of Ba/F3-BCR-ABL1-WT cells with an IC50 of 30 nM.
Catalog No. 50-003-54304 Supplier Medchemexpress LLC Supplier No. HY145887100MG
BCR-ABL1-IN-1 is an inhibitor for ABL1 (64-515) with an IC50 of 8.7 nM. It can be utilized in research concerning diseases related to the central nervous system (CNS).
  • Exhibits antiproliferative activity against mouse BaF3 cells expressing BCR-ABL with a GI50 of 69 nM and an IC50 of 75 nM.
  • Inhibits the proliferation of Ba/F3-BCR-ABL1-WT cells with an IC50 of 30 nM.