Description
- Abrasion resistant polyester knit clean room fabric laminated to absorbent polyurethane foam
- Laser cut to minimize fiber and particle release
- 10 mm thickness provides cushioned surface for optimum conformance to uneven floors
- High absorbency capacity
- Compatible with a wide range of chemicals, disinfectants and cleaners
- Autoclavable and compatible with gamma and ETO sterilization
Specifications
Specifications
| Product Type | Mop Cover |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Disposable | Single-use |
| Color-coded | No |
| Material | Knitted Polyester Microfiber |