Learn More
Sino Biological Recombinant Human AURKC/Aurora C Protein (Full Length, GST Tag)
Catalog No. 502575887
Encompass_Preferred
Recombinant full length human AURORA C was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5887 Supplier Sino Biological Supplier No. A3314G100
Recombinant full length human AURORA C was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.