Description
BCR™ Small Aluminum Mop Frame replicates the most effective way to clean a surface- wiping. Easy to use, highly effective and quick. Flat Mop Head is light and easy to maneuver, even with wet wipers attached. Clean, light and durable.
- Clean, light and durable
- High quality, anodized aluminum
- Autoclavable
- Plastic eyes for wiper attachment
Specifications
Specifications
| Product Type | Flat Mop |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Color-coded | No |
| Material | Anodized Aluminum |