Learn More
Enzo Life Sciences Breast cancer resistance protein monoclonal antibody (BXP-53) (50µg)
SDP
Supplier:  Enzo Life Sciences ALX801036C050
Encompass_Preferred
Clone: BXP-53. Isotype: IgG2a. Immunogen: Recombinant mouse BCRP (breast cancer resistance protein; MXR) (aa 221-394). Species reactivity: Human, Mouse. Specificity: Recognizes an internal epitope of BCRP. Applications: ICC, IHC (FS), IHC (PS), WB. Formulation: Liquid. In PBS, containing 0.1% BSA, and 0.02% sodium azide.
Catalog No. 50-200-9704