Learn More
Proteintech Group Inc BCRP,ABCG2 Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 272861AP150UL
Encompass_Preferred
The BCRP,ABCG2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human BCRP,ABCG2. This antibody recognizes human, mouse antigen. The BCRP,ABCG2 antibody has been validated for the following applications: IHC, WB,ELISA analysis.
Catalog No. 50-225-6459