Learn More
Siemens Healthineers BCS™ XP System, Complete
Catalog No. 23044717 Shop All Siemens Healthineers Products
Change view
Click to view available options
Throughput:
380 PT Tests/Hr., 325 Simultaneous PT/APTT Tests/Hr.
Catalog No. Throughput
23-044-717 380 PT Tests/Hr., 325 Simultaneous PT/APTT Tests/Hr.
1 options
This item is not returnable. View return policy
Catalog No. 23-044-717 Supplier Siemens Healthineers Supplier No. 10459330
This product is not orderable online. For ordering, please contact your Fisher Healthcare Sales Representative.
This item is not returnable. View return policy
The Behring Coagulation System (BCS™ XP System) performs quantitative assays of specific parameters in human citrated plasma.
The BCS™ XP System, one of the most widely used fully automated hemostasis analyzers, delivers accurate and precise results. The high-speed, random-access analyzer performs clotting, chromogenic, immunologic, and agglutination testing while allowing the consolidation of both specialty and routine reagents onto one high throughput analyzer.
  • User-friendly software and customizable interface allow for versatile user-defined configurations to meet your lab’s specific needs.
  • Automatic monitoring of reagent levels and onboard stability tracking enable personnel to focus on what’s important—critical results.
  • Broad routine and specialty testing menu, including vWF ristocetin and INNOVANCE™ ETP, allow for platform consolidation.
  • Reagent and sample probe crash sensors protect capped reagent bottles or sample tubes from damage.
  • True random-access and walk-away ability for both normal and pathological samples maximize high-volume routine workloads.
TRUSTED_SUSTAINABILITY
Specifications
Type BCS XP System
Sample Size 100 Samples (10 Racks)
Throughput 380 PT Tests/Hr., 325 Simultaneous PT/APTT Tests/Hr.
Depth (Metric) 630 mm
Height (Metric) 920 mm
Width (Metric) 1240 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov