Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ BCSA, USP, Selective Culture Media for Burkholderia Cepacia
SDP

Catalog No. 23001755 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-755 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-755 Supplier Hardy Diagnostics™ Supplier No. G09
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the selective osilation and differentiation of Burkholderia cepacia from sputum samples from cystic fibrosis patients and other clinical specimens

  • Hardy Diagnostics BCSA, USP contains peptones and sugars that supply nutrients for the growth of Burkholderia cepacia and other microorganisms
  • Crystal violet is added to inhibit growth of gram-positive organisms
  • Formulated in accordance with the U.S. Pharmacopoeia standard formula for this medium

Specifications

Appearance Slightly Opalescent
Color Reddish-orange
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Gentamicin, Vancomycin, Crystal Violet, Polymyxin B
Detects Burkholderia cepacia
Form Prepared Medium
Format Plate
Manufacturing Origin United States
Media Type Burkholderia cepacia Selective Agar (BCSA)
Product Type Agar Plate
Sterility Sterile
Target Organism Class Non-fastidious, Gram-negative Organisms
Test Type Microbial Differentiation
Fill Volume 18 mL
Formulation Casein peptone - 10.0g, Lactose - 10.0g, Sucrose - 10.0g, Sodium chloride - 5.0g, Yeast extract - 1.5g, Phenol red - 0.08g, Gentamicin - 10.0mg, Vancomycin - 2.5mg, Crystal violet - 2.0mg, Polymyxin B - 600,000U, Agar - 14.0g
For Use With (Application) Selective isolation, Microbial differentiation
Incubation Conditions 30°C to 35° C
Incubation Time 48 to 72 hr.
Material Polystyrene
No. per Pack 10
Packaging Type Sleeve, Breathable Bag
Quantity 10 Ea.
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Burkholderia cepacia
Protein Source Casein, Yeast
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.