Learn More
Enzo Life Sciences BCTC (10mg). CAS: 393514-24-4
SDP
Catalog No. 502010369
Encompass_Preferred
BCTC is a potent and selective inhibitor of capsaicin-induced (IC50=35 nM) and acid-induced (IC50=6 nM) VR1 activation. It displays analgesic activity in rat models of inflammatory and neuropathic pain. TRPV1 antagonist. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (20 mg/ml) or ethanol (15 mg/ml with warming). Long Term Storage: Ambient.
Catalog No. 50-201-0369 Supplier Enzo Life Sciences Supplier No. BMLCA2310010
BCTC is a potent and selective inhibitor of capsaicin-induced (IC50=35 nM) and acid-induced (IC50=6 nM) VR1 activation. It displays analgesic activity in rat models of inflammatory and neuropathic pain. TRPV1 antagonist. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (20 mg/ml) or ethanol (15 mg/ml with warming). Long Term Storage: Ambient.