Learn More
Abcam BCTC, inhibitor of TRPV1, 10MG
Catalog No. 502706538
Encompass_Preferred
MW 372.9 Da, Purity >98%. Potent inhibitor of TRPV1 (IC₅₀ values are 35 and 6 nM for capsaicin and acid-induced activation respectively). Inhibits TRPM8 (IC₅₀ = 0.8 μM). Reduces hyperalgesia in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6538 Supplier Abcam Supplier No. AB14151710MG
MW 372.9 Da, Purity >98%. Potent inhibitor of TRPV1 (IC₅₀ values are 35 and 6 nM for capsaicin and acid-induced activation respectively). Inhibits TRPM8 (IC₅₀ = 0.8 μM). Reduces hyperalgesia in vivo..
The product is subject to the following: Abcam Restricted Use Statement