Learn More
Innovative Research Inc Bctr-Der NEU1 Pur Lyo 25un
Catalog No. 502744629
Encompass_Preferred
Bacteria-Derived Neuraminidase Purified lyophilized 25un - Bacteria-Derived Neuraminidase Purified lyophilized from Innovative Research is useful in removing sialic acid from a variety of glycoproteins. This is a lyophilized powder. Contaminating proteolytic activity ? 0.1% using trypsin as standard. Optimized pH for the enzyme is 5.0-5.1 with little to no activity observed at pH 4.0 or above pH 8.0. Product should be protected from moisture. More Details Source Bacteria (Clostridium perfringens) Purification Chromatographically purified Storage Conditions 2-8 Degrees C
Catalog No. 50-274-4629 Supplier Innovative Research Inc Supplier No. IBCTNEUPFLY25UN
Bacteria-Derived Neuraminidase Purified lyophilized 25un - Bacteria-Derived Neuraminidase Purified lyophilized from Innovative Research is useful in removing sialic acid from a variety of glycoproteins. This is a lyophilized powder. Contaminating proteolytic activity ? 0.1% using trypsin as standard. Optimized pH for the enzyme is 5.0-5.1 with little to no activity observed at pH 4.0 or above pH 8.0. Product should be protected from moisture. More Details Source Bacteria (Clostridium perfringens) Purification Chromatographically purified Storage Conditions 2-8 Degrees C