Learn More
Cayman Chemical Bcuculn 100mg
SDP
Catalog No. 502953852
Encompass_Preferred
A competitive GABAA receptor antagonist that at 100 μM blocks spontaneously opening chloride channels in the outside-out patches from the cultured cortical neurons; reversibly blocks GABAA receptors on horizontal cells in the mouse retina (IC50 = 1.7 μM); mimics the action of epilepsy and thus used in experimental studies as a convulsant
Catalog No. 50-295-3852 Supplier Cayman Chemical Supplier No. 11727100 MG
A competitive GABAA receptor antagonist that at 100 μM blocks spontaneously opening chloride channels in the outside-out patches from the cultured cortical neurons; reversibly blocks GABAA receptors on horizontal cells in the mouse retina (IC50 = 1.7 μM); mimics the action of epilepsy and thus used in experimental studies as a convulsant