Learn More
Cayman Chemical Bicuculline, Purity: ≥98%, CAS: 485-49-4, 50 mg
SDP
Catalog No. 502952720
Encompass_Preferred
A competitive GABAA receptor antagonist that at 100 μM blocks spontaneously opening chloride channels in the outside-out patches from the cultured cortical neurons; reversibly blocks GABAA receptors on horizontal cells in the mouse retina (IC50 = 1.7 μM); mimics the action of epilepsy and thus used in experimental studies as a convulsant
Catalog No. 50-295-2720 Supplier Cayman Chemical Supplier No. 1172750 MG
A competitive GABAA receptor antagonist that at 100 μM blocks spontaneously opening chloride channels in the outside-out patches from the cultured cortical neurons; reversibly blocks GABAA receptors on horizontal cells in the mouse retina (IC50 = 1.7 μM); mimics the action of epilepsy and thus used in experimental studies as a convulsant