Learn More
Bioworld BCYE Agar (Legionella Agar), 500 g
Catalog No. 50254274
Encompass_Preferred
BCYE Agar (Legionella Agar)Commonly used in primary isolation of Legionella from environmental and clinical materialspH: 6.85Dehydrated Appearance: dark grey to black, homogeneous powderPrepared Appearance: black-green to gray-black agarSoluble in boiling waterContains (g/L):Yeast Extract: 10g Charcoal, Activated: 1.5g ACES Buffer: 10g α-Ketoglutarate: 1g Agar: 15g
Catalog No. 50-254-274 Supplier Bioworld Supplier No. 306200071
BCYE Agar (Legionella Agar)Commonly used in primary isolation of Legionella from environmental and clinical materialspH: 6.85Dehydrated Appearance: dark grey to black, homogeneous powderPrepared Appearance: black-green to gray-black agarSoluble in boiling waterContains (g/L):Yeast Extract: 10g Charcoal, Activated: 1.5g ACES Buffer: 10g α-Ketoglutarate: 1g Agar: 15g