Learn More
Medchemexpress LLC Bd-1008 dihydrobromide | 138356-09-9 | 98.1% | 463.08 g/mol | C15H24Br2Cl2N2 | 25 MG
SDP
Catalog No. 5000212230
Encompass_Preferred
BD-1008 dihydrobromide is a research-grade sigma (σ) receptor antagonist used in neuropharmacology and addiction-related studies. It is commonly applied in receptor binding assays, dopamine microdialysis, and preclinical pharmacology to probe σ receptor function and dopaminergic signaling.
  • High affinity for σ1 and σ2 receptors (Ki ≈ 2 nM and 8 nM).
  • Low affinity for D2 receptor and dopamine transporter, reducing off-target effects.
  • High purity (98.07%), suitable for research applications.
  • Available in small research pack sizes, including 25 MG.
  • Solid form with molecular weight of approximately 463.08 g/mol.
Catalog No. 50-002-12230 Supplier Medchemexpress LLC Supplier No. HY10096625MG
BD-1008 dihydrobromide is a research-grade sigma (σ) receptor antagonist used in neuropharmacology and addiction-related studies. It is commonly applied in receptor binding assays, dopamine microdialysis, and preclinical pharmacology to probe σ receptor function and dopaminergic signaling.
  • High affinity for σ1 and σ2 receptors (Ki ≈ 2 nM and 8 nM).
  • Low affinity for D2 receptor and dopamine transporter, reducing off-target effects.
  • High purity (98.07%), suitable for research applications.
  • Available in small research pack sizes, including 25 MG.
  • Solid form with molecular weight of approximately 463.08 g/mol.