Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bd-1008 dihydrobromide | 138356-09-9 | MFCD00209903 | >95.0% | 463.08 g/mol | C15H24Br2Cl2N2 | 5 MG
SDP

Catalog No. 5000212556
Encompass_Preferred

BD-1008 dihydrobromide is the dihydrobromide salt of BD-1008, a selective sigma (σ) receptor antagonist supplied as a purified research reagent. It is intended for laboratory research use to study sigma receptor pharmacology, signaling pathways, and modulation of stimulant or neurotoxic effects in preclinical models.

  • Selective sigma receptor antagonist activity
  • Dihydrobromide salt form for improved handling and solubility
  • High purity (≥95%) suitable for research applications
  • Small milligram pack sizes for preclinical use (e.g., 1-10 mg)
  • CAS number 138356-09-9 for unambiguous identification
  • Molecular weight ≈463.08 g/mol (salt form)

Catalog No. 50-002-12556 Supplier Medchemexpress LLC Supplier No. HY1009665MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BD-1008 dihydrobromide is the dihydrobromide salt of BD-1008, a selective sigma (σ) receptor antagonist supplied as a purified research reagent. It is intended for laboratory research use to study sigma receptor pharmacology, signaling pathways, and modulation of stimulant or neurotoxic effects in preclinical models.

  • Selective sigma receptor antagonist activity
  • Dihydrobromide salt form for improved handling and solubility
  • High purity (≥95%) suitable for research applications
  • Small milligram pack sizes for preclinical use (e.g., 1-10 mg)
  • CAS number 138356-09-9 for unambiguous identification
  • Molecular weight ≈463.08 g/mol (salt form)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.