Learn More
Medchemexpress LLC BD-1008 dihydrobromide | 138356-09-9 | MFCD00209903 | 98.1% | 463.08 | C15H24Br2Cl2N2 | 10 MG
SDP
Catalog No. 5000211532
Encompass_Preferred
BD-1008 dihydrobromide is a sigma receptor antagonist provided as a solid for research use. It is intended for in vitro and preclinical pharmacology studies, with characterized purity, molecular weight, and available milligram-scale pack sizes.
  • Provided as a solid suitable for storage and handling in research labs.
  • High reported purity for reliable pharmacological testing.
  • Supplied in small milligram pack sizes to support experimental dosing.
  • Characterized molecular weight and formula facilitate analytical confirmation.
  • Accompanied by safety and handling documentation for laboratory use.
Catalog No. 50-002-11532 Supplier Medchemexpress LLC Supplier No. HY10096610MG
BD-1008 dihydrobromide is a sigma receptor antagonist provided as a solid for research use. It is intended for in vitro and preclinical pharmacology studies, with characterized purity, molecular weight, and available milligram-scale pack sizes.
  • Provided as a solid suitable for storage and handling in research labs.
  • High reported purity for reliable pharmacological testing.
  • Supplied in small milligram pack sizes to support experimental dosing.
  • Characterized molecular weight and formula facilitate analytical confirmation.
  • Accompanied by safety and handling documentation for laboratory use.