Learn More
Medchemexpress LLC BD-1008 dihydrobromide | 138356-09-9 | MFCD00209903 | 98.1% | 463.08 g/mol | C15H24Br2Cl2N2 | 50 MG
SDP
Catalog No. 5000211529
Encompass_Preferred
BD-1008 dihydrobromide is a research-grade, nonselective sigma (σ) receptor antagonist provided as the dihydrobromide salt. It is supplied with reported high purity and standard physicochemical identifiers to support biochemical and pharmacological studies, enabling consistent experimental use.
  • Nonselective sigma receptor antagonist for pharmacological studies.
  • Provided as the dihydrobromide salt for stability and handling.
  • Reported purity 98.1% to support reproducible results.
  • Molecular weight 463.08 g/mol and formula C15H24Br2Cl2N2 for clear identification.
  • Available in small research pack sizes, including 50 MG, for laboratory use.
Catalog No. 50-002-11529 Supplier Medchemexpress LLC Supplier No. HY10096650MG
BD-1008 dihydrobromide is a research-grade, nonselective sigma (σ) receptor antagonist provided as the dihydrobromide salt. It is supplied with reported high purity and standard physicochemical identifiers to support biochemical and pharmacological studies, enabling consistent experimental use.
  • Nonselective sigma receptor antagonist for pharmacological studies.
  • Provided as the dihydrobromide salt for stability and handling.
  • Reported purity 98.1% to support reproducible results.
  • Molecular weight 463.08 g/mol and formula C15H24Br2Cl2N2 for clear identification.
  • Available in small research pack sizes, including 50 MG, for laboratory use.