Learn More
Cayman Chemical BD 1008hydrobromIde 1mg
SDP
Catalog No. 502894683
Encompass_Preferred
A selective antagonist of the σ1 receptor (Ki = 2 nM) that displays 4-fold selectivity for σ1 over σ2; used to modulate signaling through σ receptors by cocaine and in models of learning and memory impairment
Catalog No. 50-289-4683 Supplier Cayman Chemical Supplier No. 90013361 MG
A selective antagonist of the σ1 receptor (Ki = 2 nM) that displays 4-fold selectivity for σ1 over σ2; used to modulate signaling through σ receptors by cocaine and in models of learning and memory impairment