Learn More
Apexbio Technology LLC BD 1047 dihydrobromide 138356-21-5 50mg
SDP
Catalog No. 5000089015
Encompass_Preferred
BD 1047 dihydrobromide (CAS 138356-21-5) is a selective antagonist of the sigma-1 ( 1) receptor exhibiting Ki values of 0 93 nM for 1 and 47 nM for sigma-2 ( 2) receptors Sigma receptors distinct from opioid receptors include 1 and 2 subtypes and are implicated in modulating dopaminergic neurotransmission and drug-related behaviors In preclinical models BD 1047 dihydrobromide attenuates haloperidol- and DTG-induced dystonia and reverses cocaine-induced behavioral responses without affecting responses to natural rewards Additionally it alleviates mechanical allodynia in neuropathic pain models and modulates spinal 1 receptor and NR1 subunit expression This compound is utilized to investigate 1 receptor function in neuropharmacology and pain research
Catalog No. 50-000-89015 Supplier Apexbio Technology LLC Supplier No. B652150
BD 1047 dihydrobromide (CAS 138356-21-5) is a selective antagonist of the sigma-1 ( 1) receptor exhibiting Ki values of 0 93 nM for 1 and 47 nM for sigma-2 ( 2) receptors Sigma receptors distinct from opioid receptors include 1 and 2 subtypes and are implicated in modulating dopaminergic neurotransmission and drug-related behaviors In preclinical models BD 1047 dihydrobromide attenuates haloperidol- and DTG-induced dystonia and reverses cocaine-induced behavioral responses without affecting responses to natural rewards Additionally it alleviates mechanical allodynia in neuropathic pain models and modulates spinal 1 receptor and NR1 subunit expression This compound is utilized to investigate 1 receptor function in neuropharmacology and pain research