Learn More
Medchemexpress LLC BD-1047 dihydrobromide | 138356-21-5 | 98.45% | 437.04 | 1 ML
SDP
Catalog No. 5000391248
Encompass_Preferred
BD-1047 dihydrobromide is a selective functional antagonist of sigma receptors. It attenuates Apomorphine-induced climbing and Phencyclidine-induced head twitches. It is for research use only and not sold to patients.
  • Selective functional antagonist of sigma receptors.
  • Attenuates Apomorphine-induced climbing.
  • Attenuates Phencyclidine-induced head twitches.
  • Prevents Cutamesine from reducing cell death induced by light exposure in murine photoreceptor-derived 661w cells.
  • Attenuates Cutamesine from reducing mitochondrial damage and elevated caspase 3/7 activity.
  • Decreases APO-induced climbing behavior at 10 mg/kg in mice.
  • Counteracts the antidepressant-like effect induced by co-administration of pramipexole and sertraline.
  • Reduces the increasing expression of pNR1 and reverses Sig-1 R agonists potentiated NMDA-induced pain behavior and pNR1 immunoreactivity.
Catalog No. 50-003-91248 Supplier Medchemexpress LLC Supplier No. HY16996A10MM/1ML
BD-1047 dihydrobromide is a selective functional antagonist of sigma receptors. It attenuates Apomorphine-induced climbing and Phencyclidine-induced head twitches. It is for research use only and not sold to patients.
  • Selective functional antagonist of sigma receptors.
  • Attenuates Apomorphine-induced climbing.
  • Attenuates Phencyclidine-induced head twitches.
  • Prevents Cutamesine from reducing cell death induced by light exposure in murine photoreceptor-derived 661w cells.
  • Attenuates Cutamesine from reducing mitochondrial damage and elevated caspase 3/7 activity.
  • Decreases APO-induced climbing behavior at 10 mg/kg in mice.
  • Counteracts the antidepressant-like effect induced by co-administration of pramipexole and sertraline.
  • Reduces the increasing expression of pNR1 and reverses Sig-1 R agonists potentiated NMDA-induced pain behavior and pNR1 immunoreactivity.