Learn More
Abcam BD 1047 dihydrobromide, sigma1 receptor antagonist, 50MG
Catalog No. 502705628
Encompass_Preferred
MW 437 Da, Purity >99%. Selective σ1 receptor antagonist. Shows centrally-mediated antianalgesic, antiaddictive and antidystonic effects in vivo. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5628 Supplier Abcam Supplier No. AB14456050MG
MW 437 Da, Purity >99%. Selective σ1 receptor antagonist. Shows centrally-mediated antianalgesic, antiaddictive and antidystonic effects in vivo. Blood-brain barrier permeable.
The product is subject to the following: Abcam Restricted Use Statement