Learn More
Cayman Chemical BD 1047 (hydrobromide), Purity: ≥98%, CAS: 138356-21-5, 5 mg
SDP
Catalog No. 502993926
Encompass_Preferred
A selective antagonist of σ1 receptors (Ki = 0.9 nM in a radioligand binding assay); selective for σ1 receptors with binding affinity values greater than 10,000 nM for human recombinant dopamine, opioid, PCP, and serotonin receptors in vitro; protects against convulsions and lethality induced by cocaine and reduces cocaine-induced locomotor activity; reduces haloperidol- and DTG-induced dystonias in vivo; attenuates mechanical allodynia and microglial activation in a rat model of bone cancer pain
Catalog No. 50-299-3926 Supplier Cayman Chemical Supplier No. 229285 MG
A selective antagonist of σ1 receptors (Ki = 0.9 nM in a radioligand binding assay); selective for σ1 receptors with binding affinity values greater than 10,000 nM for human recombinant dopamine, opioid, PCP, and serotonin receptors in vitro; protects against convulsions and lethality induced by cocaine and reduces cocaine-induced locomotor activity; reduces haloperidol- and DTG-induced dystonias in vivo; attenuates mechanical allodynia and microglial activation in a rat model of bone cancer pain