Learn More
Medchemexpress LLC HY-P7136 10ug Medchemexpress, BD-2, Mouse CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP713610UG
Encompass_Preferred
Medchemexpress, HY-P7136 10ug BD-2, Mouse CAS: Recombinant Mouse beta Defensin-2 is an anti-microbial peptide that participates in the response to microbial invasion. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3111