Learn More
Description
- 7-AAD: 7-Amino-Actinomycin D
- Format: Solution
- Can be used in conjunction with phycoerythrin (PE)- and fluorescein isothiocyanate (FITC)-labelled monoclonal antibodies in 2-color analysis
- Minimal spectral overlap between 7-AAD, PE and FITC fluorescence emissions
- Detection range: far red range of the spectrum
- Storage buffer: Aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
Flow cytometry
Specifications
Specifications
| For Use With (Application) | Can be used in conjunction with Phycoerythrin (PE) -and Fluorescein Isothiocyanate (FITC) -labeled Monoclonal Antibodies in 2-color analysis |
| Quantity | 2 mL |
| Product Type | 7-AAD Staining Solution |
| Content And Storage | 4°C |
| Label Type | Other Labels or Dyes |
Research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.