Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Ac-DEVD-AMC Caspase-3 Substrate

Catalog No. BDB556449 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556449 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556449 Supplier BD Supplier No. 556449

missing translation for 'validMainframeMsg'

Synthetic tetrapeptide fluorogenic substrate for Caspase-3 (CPP32) containing the amino acid sequence of the PARP cleavage site at Asp-216

Members of the ICE/CED-3 cysteine protease family have key roles in inflammation and mammalian apoptosis. The ICE family member Caspase-3 (also known as CPP32, Yama, apopain) is activated early in apoptosis and appears to be involved in the proteolysis of several important molecules, including poly (ADP ribose) polymerase (PARP). Activated Caspase-3 cleaves PARP from its 116 kDa to an 85 kDa residual fragment. The cleavage site in PARP is C-terminal to Asp-216. The upstream sequence of the cleavage site, DEVD (Asp-Glu-Val-Asp), is utilized as a basis for the highly specific Caspase-3 substrate, Ac (N-acetyl)-DEVD-AMC (7-amino-4-methylcoumarin).

  • Format: Purified
  • Storage buffer: Lyophilized powder

Functional assay

TRUSTED_SUSTAINABILITY

Specifications

Form Lyophilized
For Use With (Application) For Caspase-3, fluorescence quantitation, used in protease assays
Quantity 1 mg
Substrate Type Caspase Substrate
Includes Amino acid sequence of the PARP cleavage site at Asp 216
Molecular Weight (g/mol) 694
Storage Requirements -20°C
Substrate Quantity 1mg

Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.