Learn More
BD Cell Analysis 3P C6 Plus 1-year Maintenance Kit
Catalog No. NC1730076
Encompass
Contains (12) Peristaltic Pump Tubing, (6) Sheath Bottle Filters and (6) In-Line Sheath Filters.
Catalog No. NC1730076 Supplier BD Cell Analysis 3P Supplier No. 661331
Contains (12) Peristaltic Pump Tubing, (6) Sheath Bottle Filters and (6) In-Line Sheath Filters.