Learn More
BD Acridine Orange Stain
Catalog No. B12536 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
250 mL
4 x 250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12536 250 mL
B12537 4 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12536 Supplier BD Supplier No. 212536
Add to Cart
Add to Cart
For detecting microorganisms in direct smears by fluorescence
  • Especially beneficial in screening normally sterile specimens where few organisms may be present
  • Bacteria stain bright orange; human cells, tissue debris and proteinaceous material stain pale green to yellow
TRUSTED_SUSTAINABILITY
Specifications
Quantity 250 mL
Certifications/Compliance CE
For Use With (Application) Recommended for use in the fluorescent microscopic detection of microorganisms in direct smears prepared from clinical and non clinical materials
Stain Type Acridine Orange
Volume (Metric) 250 mL
Product Type Acridine Orange Stain