Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD AEC Substrate Kit

Catalog No. BDB551015 Shop All BD Biosciences Products
Change view
Click to view available options
Chromagen Quantity:
1.5 mL
3 mL
Includes:
AEC buffer and AEC chromogen
Plates, AEC buffer, chromogen
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chromagen Quantity Includes
BDB551015 1.5 mL AEC buffer and AEC chromogen
BDB551951 3 mL Plates, AEC buffer, chromogen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Contains Dimethylsulfoxide, Methanol, Urea Peroxide, 3-Amino-9-Ethylcarbazole, and N, N,Dimethylformamide which are considered as harmful.
Catalog No. BDB551015 Supplier BD Supplier No. 551015
Add to Cart
Add to Cart
Contains Dimethylsulfoxide, Methanol, Urea Peroxide, 3-Amino-9-Ethylcarbazole, and N, N,Dimethylformamide which are considered as harmful.

Includes

AEC Buffer and AEC Chromogen

Substrate for peroxidase and is a widely used chromogen for immunohistochemical staining and immunoblotting

Immunohistochemistry, ELISPOT

TRUSTED_SUSTAINABILITY

Specifications

Chromagen Quantity 1.5 mL
Chromagen Type AEC
For Use With (Application) Immunohistochemical staining and immunoblotting, ELISPOT
Substrate Type AEC Substrate
Includes AEC buffer and AEC chromogen
pH 7.2
Storage Requirements 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.