Learn More
BD Affirm™ VPIII Ambient Temperature Transport System (ATTS)
Catalog No. 23025604 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 Pk.
Catalog No. Quantity
23-025-604 100 Pk.
1 options
This item is not returnable. View return policy
Catalog No. 23-025-604 Supplier BD Supplier No. 446255
 Restricted Item
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Includes
sterile swab, sample collection tube and cap, dropper, and patient labels
Sterile, ready-to-use system for collection, transport, and preservation of vaginal specimens
For use only with BD Affirm VPIII Microbial Identification Test (23-025-603)
  • Stabilizes nucleic acid of Candida species, Gardnerella vaginalis, and Trichomonas vaginalis during specimen transport at ambient temperature (15-30°C) for up to 72 hours
  • Use only with those specimens where transport times are expected to exceed one hour at ambient temperature (15-30°C) or four hours at refrigerated temperatures (2-8°C)
TRUSTED_SUSTAINABILITY
Specifications
Product Type VPIII Ambient Temperature Transport System
Includes Sterile swab, sample collection tube and cap, dropper, and patient labels
For Use With (Application) For collection and extended transport of vaginal specimens
For Use With (Equipment) BD Affirm™ VPIII microbial identification system
Quantity 100 Pk.